missing translation for 'onlineSavingsMsg'
Learn More

Ferrous Chloride Tetrahydrate, Reagent (Iron (II) Chloride Tetrahydrate), Reagents

Catalog No. 01337964
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337964 100 g
01337963 2.5 kg
01337962 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01337964 Supplier Labchem Inc Supplier No. C1206500100C2

Ferrous Chloride Tetrahydrate, Reagent (Iron (II) Chloride Tetrahydrate), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 13478-10-9
Molecular Formula Cl2FeH8O4
Molecular Weight (g/mol) 198.81
InChI Key WSSMOXHYUFMBLS-UHFFFAOYSA-L
Synonym Iron(II) Chloride Tetrahydrate
IUPAC Name tetrahydroxyironbis(ylium) dichloride
SMILES O.O.O.O.[Cl-].[Cl-].[Fe++]

Specifications

Boiling Point Sublimes
Melting Point 670°C to 674°C
Color Green to Blue-Green
Physical Form Solid
Quantity 100 g
Percent Purity ≥ 98%
Grade Reagent
Packaging Glass Bottle
Specific Gravity 1.93
Chemical Name or Material Ferrous Chloride Tetrahydrate, Reagent (Iron (II) Chloride Tetrahydrate)