missing translation for 'onlineSavingsMsg'
Learn More

Ferrous Chloride Tetrahydrate, (Crystalline),MP Biomedicals

Catalog No. ICN15804691
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15804691 1 kg
ICN15804683 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15804691 Supplier MP Biomedicals Inc Supplier No. 0215804691

Chemical Identifiers

CAS 13478-10-9
Molecular Formula Cl2FeH8O4
Molecular Weight (g/mol) 198.805
InChI Key WSSMOXHYUFMBLS-UHFFFAOYSA-L
Synonym ferrous chloride tetrahydrate, iron ii chloride tetrahydrate, iron dichloride tetrahydrate, cl2fe.4h2o, fecl2.4h2o, iron chloride, tetrahydrate, dichloroiron-water 1/4, iron ii chloride tetra hydrate, ae>> nciu eae r masculineiii
PubChem CID 16211588
ChEBI CHEBI:86249
IUPAC Name dichloroiron;tetrahydrate
SMILES O.O.O.O.Cl[Fe]Cl

Specifications

Physical Form Crystalline
Quantity 1 kg
Formula Weight 198.8
Chemical Name or Material Ferrous Chloride Tetrahydrate
Recommended Storage Room Temperature