missing translation for 'onlineSavingsMsg'
Learn More

Ferrous Chloride, Crystal, Technical, Spectrum™ Chemical

$435.43 - $19446.11

Chemical Identifiers

CAS 13478-10-9
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18610570
View Documents
Spectrum Chemical Mfg Cor
F1056125GM
125 g Amber Glass Bottle
Each for $435.43
Only null left
 
18610571
View Documents
Spectrum Chemical Mfg Cor
F1056500GM
500 g Amber Glass Bottle
Each for $996.86
Only null left
 
18610572
View Documents
Spectrum Chemical Mfg Cor
F105625KG
2.5 kg Amber Glass Bottle
Each for $3,862.29
Only null left
 
18610573
View Documents
Spectrum Chemical Mfg Cor
F105612KG
12 kg Poly Pail
Each for $7,997.11
Only null left
 
18610574
View Documents
Spectrum Chemical Mfg Cor
F105650KGBL
50 kg Fiber Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

13478-10-9

Specifications

13478-10-9
125 g
Amber Glass Bottle
1
Technical
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.