missing translation for 'onlineSavingsMsg'
Learn More

Ferrous Chloride, Crystal, 99%, Spectrum™ Chemical

Catalog No. p-7165554
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610564 25 g Amber Glass Bottle
18610565 125 g Amber Glass Bottle
18610568 500 g Amber Glass Bottle
18610567 2.5 kg Amber Glass Bottle
18610566 12 kg Poly Pail
18610569 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18610564 Supplier Spectrum Chemical Mfg Cor Supplier No. F105525GM
Only null left

Chemical Identifiers

CAS 13478-10-9
Molecular Formula Cl2FeH8O4
Molecular Weight (g/mol) 198.81
InChI Key WSSMOXHYUFMBLS-UHFFFAOYSA-L
IUPAC Name tetrahydroxyironbis(ylium) dichloride
SMILES O.O.O.O.[Cl-].[Cl-].[Fe++]

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity 99%
Grade Reagent
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.