missing translation for 'onlineSavingsMsg'
Learn More

Ferrous Ammonium Sulfate Hexahydrate (Crystalline/Certified ACS), Fisher Chemical™

Catalog No. p-32041
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
I77500 500 g Poly Bottle
I77212 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I77500 Supplier Thermo Fisher Scientific Supplier No. I77 500
Only null left

Chemical Identifiers

CAS 7783-85-9
Molecular Formula FeH20N2O14S2
Molecular Weight (g/mol) 392.13
MDL Number MFCD00150530
InChI Key MQLVWQSVRZVNIP-UHFFFAOYSA-L
Synonym Mohr Salt, Iron (II) Ammonium Sulfate Hexahydrate
IUPAC Name λ²-iron(2+) diammonium hexahydrate disulfate
SMILES [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Melting Point 100°C
Color Green
pH 3,5
Physical Form Solid
Quantity 500 g
Assay Percent Range 98.5 to 101.5 %
Linear Formula Fe(NH4)2(SO4)2·6H2O
Formula Weight 392.14
Percent Purity 98.5 to 101.5%
Grade Certified ACS
Identification Pass test
Packaging Poly Bottle
Chemical Name or Material Ferrous Ammonium Sulfate Hexahydrate
Show More Show Less
Hazard Category Signal Word: WARNING!
Precautionary Statement Emergency Overview
Irritating to eyes, respiratory system and skin.
NFPA
Health: 1
Flammability: