missing translation for 'onlineSavingsMsg'
Learn More

Ferrous Ammonium Sulfate Hexahydrate, 98.5-101.5%, (ACS Reagent Grade), MP Biomedicals

Catalog No. p-3547114
Change view
Click to view available options
Quantity:
500 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15252390 500 g
ICN15252391 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15252390 Supplier MP Biomedicals Inc Supplier No. 0215252390
Only null left

Chemical Identifiers

CAS 7783-85-9
Molecular Weight (g/mol) 392.13
MDL Number MFCD00150530
InChI Key MQLVWQSVRZVNIP-UHFFFAOYSA-L
IUPAC Name λ²-iron(2+) diammonium hexahydrate disulfate
SMILES [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Quantity 500 g
Assay Percent Range 98.5-101.5%
Molecular Formula Fe[NH4]2[SO4]2 · 6H2O
Formula Weight 392.1
Grade ACS Reagent
Chemical Name or Material Ferrous Ammonium Sulfate Hexahydrate
Recommended Storage Room Temperature