missing translation for 'onlineSavingsMsg'
Learn More

Ferrous Ammonium Sulfate, Hexahydrate, 98.5 to 101.5%, ACS, MilliporeSigma™

Catalog No. MFX02451
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MFX02451 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MFX02451 Supplier Technidata Supplier No. FX02451

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ACS reagent grade ammonium ferrous sulfate suitable for analytical titration and general redox reactions.

Chemical Identifiers

CAS 7783-85-9
Molecular Formula FeH20N2O14S2
Molecular Weight (g/mol) 392.13
MDL Number MFCD00150530
InChI Key MQLVWQSVRZVNIP-UHFFFAOYSA-L
Synonym Ferrous ammonium sulfate, Mohr's salt, Iron(II) ammonium sulfate
IUPAC Name λ²-iron(2+) diammonium hexahydrate disulfate
SMILES [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Color Blue-Green
Physical Form Crystals
Quantity 500 g
Assay Percent Range 98.5 to 101.5%
Linear Formula (NH4)2Fe(SO4)2 · 6H2O
Formula Weight 392.14
Percent Purity 98.5% to 101.5%
Grade ACS
Density 900kg/m3
Chemical Name or Material Ferrous Ammonium Sulfate, Hexahydrate, EMSURE™, For Analysis
Recommended Storage Store at +15°C to 25°C