missing translation for 'onlineSavingsMsg'
Learn More

Ferrous Ammonium Sulfate, Certified, 0.100N ±0.006N (0.1M), LabChem™

Supplier:  LabChem LC145004

 View more versions of this product

Catalog No. LC145004


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Ferrous Ammonium Sulfate
Green
Liquid
FeH20N2O14S2
MFCD00150530
Ammonium Iron (II) Sulfate, Iron (II) Ammonium Sulfate Hexahydrate, Iron(II) ammonium sulfate
MQLVWQSVRZVNIP-UHFFFAOYSA-L
λ²-iron(2+) diammonium hexahydrate disulfate
115148
Certified
Passes Test
Gaseous Ammonia; Sulfur compounds; Thermal decomposition generates: Corrosive vapors
7783-85-9,7732-18-5,7664-93-9
3.92,94.08,2
4 L
Fe(NH4)2(SO4)2·6H2O
UN3264
Soluble in water
[NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
392.13
392.14
0.100N ±0.006N (0.1M)
Amber Glass

Chemical Identifiers

7783-85-9
392.13
MQLVWQSVRZVNIP-UHFFFAOYSA-L
115148
[NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
FeH20N2O14S2
MFCD00150530
Ammonium Iron (II) Sulfate, Iron (II) Ammonium Sulfate Hexahydrate, Iron(II) ammonium sulfate
λ²-iron(2+) diammonium hexahydrate disulfate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

Danger

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions