missing translation for 'onlineSavingsMsg'
Learn More

Ferrous Ammonium Sulfate, 6-Hydrate, Fine Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog No. p-5390756
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
Packaging:
Poly Bottle
Wide Mouth Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02002884 500 g Wide Mouth Amber Glass Bottle
02002882 1 kg Poly Bottle
02002883 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02002884 Supplier Avantor Supplier No. 205401
Only null left

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 7783-85-9
Molecular Weight (g/mol) 392.13
MDL Number MFCD00150530
InChI Key MQLVWQSVRZVNIP-UHFFFAOYSA-L
IUPAC Name λ²-iron(2+) diammonium hexahydrate disulfate
SMILES [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Physical Form Fine Crystals
Quantity 500 g
Molecular Formula Fe(NH4)2(SO4)2·6H2O
Formula Weight 392.13
Grade BAKER ANALYZED™ ACS Reagent
Packaging Wide Mouth Amber Glass Bottle
Chemical Name or Material Ferrous Ammonium Sulfate, 6-Hydrate