missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferrous Ammonium Sulfate, 6-Hydrate, Fine Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Specifications
Specifications
| Physical Form | Fine Crystals |
| Quantity | 500 g |
| Molecular Formula | Fe(NH4)2(SO4)2·6H2O |
| Formula Weight | 392.13 |
| Percent Purity | 98.5 to 101.5% |
| Grade | BAKER ANALYZED™ ACS Reagent |
| Packaging | Wide Mouth Amber Glass Bottle |
| Chemical Name or Material | Ferrous Ammonium Sulfate, 6-Hydrate |