missing translation for 'onlineSavingsMsg'
Learn More

Ferrous Ammonium Sulfate, 0.1N Mohr's Salt Solution, Reagents

Catalog No. 01334709
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 L
20 L
4 L
500 mL
Catalog No. Quantity
01334709 4 L
01334711 1 L
01334710 20 L
01334708 500 mL
4 options

Catalog No. 01334709

Supplier: Labchem Inc CM1297004A

Only null left

Ferrous Ammonium Sulfate, 0.1N Mohr's Salt Solution, Requires Periodic Restandardization, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Specifications

Chemical Name or Material Ferrous Ammonium Sulfate, 0.1N Mohr's Salt Solution, Requires Periodic Restandardization
Boiling Point 100°C
Color Light Green
CAS 7732-18-5, 7664-93-9
pH <1
Physical Form Liquid
Quantity 4 L
Synonym Iron(II) Ammonium Sulfate
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.05
Show More Show Less