missing translation for 'onlineSavingsMsg'
Learn More

Ferrous Ammonium Sulfate, 0.100 N (N/10), 0.100 M (M/10), Ricca Chemical

Catalog No. 31455
Change view
Click to view available options
Package: Cubitainer; Size: 20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
31455 Package: Cubitainer; Size: 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 31455 Supplier Ricca Chemical Company Supplier No. 31455

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 7783-85-9, 7732-18-5, 7664-93-9
CAS Min % 3.57, 91.0, 3.35
CAS Max % 3.72, 95.0, 3.49
Molecular Formula FeH20N2O14S2
Synonym Ammonium Iron (II) Sulfate, Iron (II) Ammonium Sulfate Hexahydrate, Iron(II) ammonium sulfate
InChI Key SHHFGSWBFZVUNE-UHFFFAOYSA-L
SMILES [NH4+].[NH4+].O.O.O.O.O.O.[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
IUPAC Name iron(3+) diammonium hexahydrate disulfate
Molecular Weight (g/mol) 392.12
PubChem CID 115148  
Packaging Cubitainer
Quantity 20 L
Concentration or Composition Notes 0.100Normal,0.100Molar
Chemical Name or Material Ferrous Ammonium Sulfate
Show More Show Less