missing translation for 'onlineSavingsMsg'
Learn More

Ferroin indicator solution, Solstice

Catalog No. 60046355
Change view
Click to view available options
Quantity:
100 mL
500 mL
6 x 100 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60046355 6 x 100 mL
6002973 100 mL
6002974 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60046355 Supplier Solstice Supplier No. 345316X100ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Redox Indicator, Reag. Ph. Eur., E0 in sulfuric acid 1 mol/l = +1,06 volt

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 14634-91-4
Molecular Formula C36H24FeN6O4S
Molecular Weight (g/mol) 692.53
MDL Number MFCD00036428
InChI Key CIWXFRVOSDNDJZ-UHFFFAOYSA-L
Synonym ferroin, ferroin indicator solution, iron 2+ , tris 1,10-phenanthroline-.kappa.n1,.kappa.n10-, oc-6-11-, sulfate 1:1, ferroin sulfate, ferroin solution, 1,10-phenanthroline ferrous sulfate, tris 1,10-phenanthroline-n1,n10 iron sulphate, ferrous 1,10-phenanthroline sulfate, ferroin indicator solution, 25 mmol/l, iron 2+ , tris 1,10-phenanthroline-kappan1,kappan10-, oc-6-11-, sulfate 1:1
PubChem CID 84567
SMILES [Fe++].[O-]S([O-])(=O)=O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1

Specifications

Quantity 6 x 100 mL
Linear Formula [Fe(C12H8N2)3]SO4
UN Number 3082
IUPAC Name iron(2+);1,10-phenanthroline;sulfate
Formula Weight 692.52
Grade Reagent Ph. Eur.
Chemical Name or Material Ferroin indicator solution
DOTInformation 9 / PGIII