missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferroin Indicator, Macron Fine Chemicals™
macron
$40792.33 - $40792.33
Specifications
| Chemical Name or Material | Ferroin Indicator |
|---|---|
| Color | Red |
| Physical Form | Solution |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
MKH119500
|
Avantor
H11903 |
500 mL |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Specifications
| Ferroin Indicator | |
| Red | |
| Solution |