missing translation for 'onlineSavingsMsg'
Learn More

Ferroin Indicator, 0.025M, Solution, Spectrum™ Chemical

Catalog No. 18610549
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18610549 100 mL
18610550 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18610549 Supplier Spectrum Chemical Mfg Cor Supplier No. F157100ML
Get Up to $500 in Chemicals with Purchase
View all promotions

Specifications

Chemical Name or Material Ferroin Indicator Solution
CAS 7782-63-0, 5144-89-8, 7732-18-5
Concentration 0.025 M
Grade Chemical Solution
Packaging Amber Glass Bottle
Color Dark Red
Quantity 100 mL

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.