missing translation for 'onlineSavingsMsg'
Learn More

Ferroin Indicator, 0.025 M Phenanthroline Ferrous Sulfate Complex, Ricca Chemical

Catalog No. p-3251680
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
314016 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 314016 Supplier Ricca Chemical Company Supplier No. 314016
Only null left

Specifications

Chemical Name or Material Ferroin Indicator
Name Note Phenanthroline Ferrous Sulfate Complex
CAS 7732-18-5, 5144-89-8, 7782-63-0
CAS Min % 96.0, 1.43, 0.67
CAS Max % 100.0, 1.48, 0.69
Concentration 0.025Molar
Percent Purity 0.025 M
Grade Indicator
Packaging Amber Glass Bottle
Color Dark Red
Quantity 500 mL
Physical Form Liquid
Show More Show Less