missing translation for 'onlineSavingsMsg'
Learn More

(Ferrocenylmethyl)trimethylammonium Bromide 95.0+%, TCI America™

Catalog No. p-7129517
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F01661G 1 g
F01665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F01661G Supplier TCI America Supplier No. F01661G
Only null left

Chemical Identifiers

CAS 106157-30-6
Molecular Formula C14H20BrFeN
MDL Number MFCD00059981

Specifications

Quantity 1 g
Formula Weight 338.07
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (Ferrocenylmethyl)trimethylammonium Bromide