missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferroceneboronic Acid (contains varying amounts of Anhydride) 98.0+%, TCI America™
$96.94 - $477.19
Chemical Identifiers
| CAS | 12152-94-2 |
|---|---|
| Molecular Formula | C10H11BFeO2 |
| MDL Number | MFCD00059074 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
F0280100MG
|
TCI America
F0280100MG |
100 mg |
Each for $96.94
|
|
|||||
|
F02801G
|
TCI America
F02801G |
1 g |
Each for $477.19
|
|
|||||
Chemical Identifiers
| 12152-94-2 | |
| C10H11BFeO2 | |
| MFCD00059074 |
Specifications
| 12152-94-2 | |
| MFCD00059074 | |
| 229.85 | |
| Crystalline Powder |
| C10H11BFeO2 | |
| 100 mg | |
| ≥98.0% (HPLC) | |
| Ferroceneboronic Acid (contains varying amounts of Anhydride) [Cyclic boronating reagent for GC/MS] |
Safety and Handling
TSCA : No