missing translation for 'onlineSavingsMsg'
Learn More

Ferroceneboronic Acid (contains varying amounts of Anhydride) 98.0+%, TCI America™

Catalog No. F02801G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F02801G 1 g
F0280100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F02801G Supplier TCI America Supplier No. F02801G
Only null left

Chemical Identifiers

CAS 12152-94-2
Molecular Formula C10H11BFeO2
MDL Number MFCD00059074

Specifications

Quantity 1 g
Formula Weight 229.85
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Ferroceneboronic Acid (contains varying amounts of Anhydride) [Cyclic boronating reagent for GC/MS]