missing translation for 'onlineSavingsMsg'
Learn More

Ferric Sulfate, Technical, Spectrum™ Chemical

Catalog No. p-7157422
Change view
Click to view available options
Quantity:
500 g
2.5 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602683 500 g Amber Glass Bottle
18602684 2.5 kg Amber Glass Bottle
18602685 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18602683 Supplier Spectrum Chemical Mfg Cor Supplier No. F1047500GM
Only null left

F1047, 15244-10-7, Fe2(SO4)3·xH2O

Spectrum™ Chemical Ferric Sulfate, Technical is used to prepare ferric subsulfate solutions. Technical grade products supplied by Spectrum are indicative of a grade suitable for general industrial use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 15244-10-7
Molecular Formula Fe2H2O13S3
Molecular Weight (g/mol) 417.87
InChI Key VXWSFRMTBJZULV-UHFFFAOYSA-H
IUPAC Name diiron(3+) hydrate trisulfate
SMILES O.[Fe+3].[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

CAS Max % 100%
Quantity 500 g
Grade Technical
Packaging Amber Glass Bottle