missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferric Sulfate, n-Hydrate, BAKER ANALYZED™ Reagent, J.T. Baker™
Specifications
Specifications
| CAS | 15244-10-7 |
| Quantity | 2.5 kg |
| Molecular Formula | Fe2H2O13S3 |
| InChI Key | VXWSFRMTBJZULV-UHFFFAOYSA-H |
| SMILES | O.[Fe+3].[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O |
| IUPAC Name | diiron(3+) hydrate trisulfate |
| Molecular Weight (g/mol) | 417.87 |
| PubChem CID | 167265 |
| Grade | BAKER ANALYZED™ Reagent |
| Packaging | Poly Bottle |
| Show More |