missing translation for 'onlineSavingsMsg'
Learn More

Ferric Sulfate Hydrate, Reagents

Catalog No. 01337970
Change view
Click to view available options
Quantity: 2.5 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01337970 Quantity: 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01337970 Supplier Labchem Inc Supplier No. C12052502.5KG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ferric Sulfate Hydrate, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 15244-10-7
Molecular Formula Fe2H2O13S3
Molecular Weight (g/mol) 417.87
InChI Key VXWSFRMTBJZULV-UHFFFAOYSA-H
Synonym Iron(III) Sulfate
IUPAC Name diiron(3+) hydrate trisulfate
SMILES O.[Fe+3].[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Boiling Point 330°C
Color Light Yellow
Physical Form Solid
Quantity 2.5 kg
Grade Reagent
Packaging Poly Bottle
Specific Gravity 3.097
Chemical Name or Material Ferric Sulfate Hydrate