missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferric Sulfate AR Powder, Macron Fine Chemicals™
macron
Chemical Identifiers
| CAS | 15244-10-7 |
|---|---|
| Molecular Formula | Fe2H2O13S3 |
| Molecular Weight (g/mol) | 417.873 |
| InChI Key | VXWSFRMTBJZULV-UHFFFAOYSA-H |
| PubChem CID | 167265 |
| IUPAC Name | iron(3+);trisulfate;hydrate |
| SMILES | O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3].[Fe+3] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
MK5036212
|
Avantor Macron
503605 |
2.5kg | STAKMOR |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 15244-10-7 | |
| 417.873 | |
| 167265 | |
| O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3].[Fe+3] |
| Fe2H2O13S3 | |
| VXWSFRMTBJZULV-UHFFFAOYSA-H | |
| iron(3+);trisulfate;hydrate |
Specifications
| 15244-10-7 | |
| VXWSFRMTBJZULV-UHFFFAOYSA-H | |
| iron(3+);trisulfate;hydrate | |
| 167265 |
| Fe2H2O13S3 | |
| O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3].[Fe+3] | |
| 417.873 | |
| Ferric Sulfate |