missing translation for 'onlineSavingsMsg'
Learn More

Ferric Subsulfate Solution, USP, 20-22 g/100 mL, Spectrum™ Chemical

F1305, 1310-45-8, Fe4(OH)2(SO4)5

$1250.14 - $31430.46

Chemical Identifiers

CAS 1310-45-8
Molecular Formula Fe4H2O22S5
Molecular Weight (g/mol) 737.67
InChI Key GDPKWKCLDUOTMP-UHFFFAOYSA-B
IUPAC Name tetrairon(3+) dihydroxide pentasulfate
SMILES [OH-].[OH-].[Fe+3].[Fe+3].[Fe+3].[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18602680
View Documents
Spectrum Chemical Mfg Cor
F1305500ML
500 mL Amber Glass Bottle
Each for $1,250.14
Only null left
 
18602681
View Documents
Spectrum Chemical Mfg Cor
F1305500MLCS6
6 x 500 mL Amber Glass Bottle
Case of 6 Each for $4,880.43
Only null left
 
18602682
View Documents
Spectrum Chemical Mfg Cor
F1305200LTBL
200 L Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Spectrum™ Chemical Ferric Subsulfate Solution, USP All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications

Chemical Identifiers

1310-45-8
737.67
tetrairon(3+) dihydroxide pentasulfate
Fe4H2O22S5
GDPKWKCLDUOTMP-UHFFFAOYSA-B
[OH-].[OH-].[Fe+3].[Fe+3].[Fe+3].[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

1310-45-8
500 mL
GDPKWKCLDUOTMP-UHFFFAOYSA-B
tetrairon(3+) dihydroxide pentasulfate
USP
Amber Glass Bottle
100%
Fe4H2O22S5
[OH-].[OH-].[Fe+3].[Fe+3].[Fe+3].[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
737.67
20-22 g/100 mL
Videos
SDS
Documents

Documents