missing translation for 'onlineSavingsMsg'
Learn More

Ferric Nitrate Solution, Stock, 202 g/L in dilute Nitric Acid, Ricca Chemical

Supplier:  Ricca Chemical Company 313432

Catalog No. 3134-32

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

Ferric Nitrate Solution
7697-37-2,7782-61-8,7732-18-5
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
[N+](=O)(O)[O-]
63.012
CHEBI:48107
Dilute Nitric Acid
202g/L
Stock Solution
HNO3
GRYLNZFGIOXLOG-UHFFFAOYSA-N
nitric acid
944
Amber Poly Bottle
1 L
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions