missing translation for 'onlineSavingsMsg'
Learn More

Ferric Nitrate Nonahydrate (Crystalline/Certified ACS), Fisher Chemical™

Catalog No. p-32085
Change view
Click to view available options
Quantity:
100 g
500 g
3 kg
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
I110100 100 g Glass Bottle
I110500 500 g Glass Bottle
I1103 3 kg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I110100 Supplier Thermo Fisher Scientific Supplier No. I110100
Only null left

Chemical Identifiers

CAS 7782-61-8
Molecular Formula FeH18N3O18
Molecular Weight (g/mol) 403.99
MDL Number MFCD00149708
InChI Key SZQUEWJRBJDHSM-UHFFFAOYSA-N
Synonym ferric nitrate nonahydrate, iron iii nitrate nonahydrate, unii-vmx4uop3vn, ferric nitrate, nonahydrate, iron iii nitrate, vmx4uop3vn, iron, reference standard solution, ferricnitratenonahydrate, nitric acid, iron 3+ salt, nonahydrate, iron iii nitrate hydrate
PubChem CID 16211566
IUPAC Name iron(3+) nonahydrate trinitrate
SMILES O.O.O.O.O.O.O.O.O.[Fe+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 47°C
Color Purple
Physical Form Solid
Quantity 100 g
Assay Percent Range 98.0 to 101.0 %
Linear Formula Fe(NO3)3·9H2O
UN Number 1466
Formula Weight 404
Percent Purity 98.0 to 101.0%
Grade Certified ACS
Identification Pass test
Packaging Glass Bottle
Vapor Pressure Negligible
Decomposition Information <100°C
Density 1.680g/cm³
Chemical Name or Material Ferric Nitrate Nonahydrate
Show More Show Less
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Oxidizer: Contact with combustible/organic material may cause fire. Hygroscopic. Use personal protective equipment. Keep away from clothing and other combustible materials. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:2
Flammability:1
Instability:2
DOTInformation DOT Class 5.1, : Oxidizer