missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferric Nitrate, 9-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
$4996.00 - $4996.00
Chemical Identifiers
| CAS | 7782-61-8 |
|---|---|
| Molecular Formula | FeH18N3O18 |
| Molecular Weight (g/mol) | 403.99 |
| MDL Number | MFCD00149708 |
| InChI Key | SZQUEWJRBJDHSM-UHFFFAOYSA-N |
| Synonym | ferric nitrate nonahydrate, iron iii nitrate nonahydrate, unii-vmx4uop3vn, ferric nitrate, nonahydrate, iron iii nitrate, vmx4uop3vn, iron, reference standard solution, ferricnitratenonahydrate, nitric acid, iron 3+ salt, nonahydrate, iron iii nitrate hydrate |
| PubChem CID | 16211566 |
Chemical Identifiers
| 7782-61-8 | |
| 403.99 | |
| SZQUEWJRBJDHSM-UHFFFAOYSA-N | |
| 16211566 |
| FeH18N3O18 | |
| MFCD00149708 | |
| ferric nitrate nonahydrate, iron iii nitrate nonahydrate, unii-vmx4uop3vn, ferric nitrate, nonahydrate, iron iii nitrate, vmx4uop3vn, iron, reference standard solution, ferricnitratenonahydrate, nitric acid, iron 3+ salt, nonahydrate, iron iii nitrate hydrate |
Specifications
| 7782-61-8 | |
| 500 g | |
| MFCD00149708 | |
| SZQUEWJRBJDHSM-UHFFFAOYSA-N | |
| iron(3+);trinitrate;nonahydrate | |
| 16211566 | |
| BAKER ANALYZED™ ACS Reagent | |
| 1L = 1.68kg |
| Crystals | |
| FeH18N3O18 | |
| ferric nitrate nonahydrate, iron iii nitrate nonahydrate, unii-vmx4uop3vn, ferric nitrate, nonahydrate, iron iii nitrate, vmx4uop3vn, iron, reference standard solution, ferricnitratenonahydrate, nitric acid, iron 3+ salt, nonahydrate, iron iii nitrate hydrate | |
| [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Fe+3] | |
| 403.99 | |
| 404 | |
| Poly Bottle | |
| Ferric Nitrate, 9-Hydrate |