missing translation for 'onlineSavingsMsg'
Learn More

Ferric Chloride Solution, Fisher Chemical™

Catalog No. SF9000250
Change view
Click to view available options
Package: Amber Glass; Size: 250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
SF9000250 Package: Amber Glass; Size: 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SF9000250 Supplier Ricca Chemical Company Supplier No. SF9000250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 10025-77-1, 7732-18-5, 7647-01-0
CAS Min % 0.97, 97.0, 0.24
CAS Max % 1.01, 100.0, 0.25
Molecular Formula Cl3FeH12O6
Synonym Ferric chloride hexahydrate, Iron Trichloride, Perchlorure de Fer
InChI Key NQXWGWZJXJUMQB-UHFFFAOYSA-K
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]
IUPAC Name hexahydroxyirontris(ylium) trichloride
Molecular Weight (g/mol) 270.29
PubChem CID 24380  
ChEBI CHEBI:30808
Packaging Amber Glass Bottle
Methods For AOAC Method 937.05
Quantity 250 mL
Chemical Name or Material Ferric Chloride Solution
Show More Show Less
Safety handling Check Alert storage code Red
DOTInformation DOT Class 8, : Corrosive