missing translation for 'onlineSavingsMsg'
Learn More

Ferric Chloride Oxidizing Solution, 1% (w/v), with 1.6% (w/v) Sulfamic Acid, Ricca Chemical

Catalog No. 311332
Change view
Click to view available options
Package: Natural Poly; Size: 1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
311332 Package: Natural Poly; Size: 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 311332 Supplier Ricca Chemical Company Supplier No. 311332

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For Cyanide Determination

Specifications

CAS 10025-77-1, 5329-14-6, 7732-18-5
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous,with 1.6% (w/v) Sulfamic Acid
Quantity 1 L
Concentration or Composition Notes 1% (w/v)
Chemical Name or Material Ferric Chloride Oxidizing Solution