missing translation for 'onlineSavingsMsg'
Learn More

Ferric Chloride, 1% (w/v), Ricca Chemical

Supplier:  Ricca Chemical Company R31080001C

 View more versions of this product

Catalog No. 01-213-130


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Approximately 100°C
Approximately 0°C
7732-18-5,10025-77-1,7647-01-0,10025-77-1,7732-18-5
Liquid
0.09%,0.99%,98.91%
Laboratory
Ferric Chloride Solution
Dark Yellow-Orange
0.09%,0.99%,98.91%
1 L
1% W/V
1 L Amber Glass Bottle

Chemical Identifiers

7732-18-5
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 12 Months

SDS
Documents

Documents

Promotions

Promotions