missing translation for 'onlineSavingsMsg'
Learn More

Ferric Ammonium Sulfate (Ferric Alum), Saturated, Ricca Chemical

Catalog No. 30704
Change view
Click to view available options
Package: Natural Poly; Size: 120 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
30704 Package: Natural Poly; Size: 120 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 30704 Supplier Ricca Chemical Company Supplier No. 30704

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 7783-83-7, 7732-18-5
CAS Min % 66.0, 32.24
CAS Max % 68.0, 33.56
Molecular Formula FeH28NO20S2
Synonym ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100g
InChI Key LCPUDZUWZDSKMX-UHFFFAOYSA-K
SMILES [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3]
IUPAC Name azanium;iron(3+);disulfate;dodecahydrate
Molecular Weight (g/mol) 482.176
PubChem CID 197097
Packaging Natural Poly Bottle
Solvent or Matrix Saturated Aqueous Solution
Quantity 120 mL
Chemical Name or Material Ferric Ammonium Sulfate, Ferric Alum
Show More Show Less