missing translation for 'onlineSavingsMsg'
Learn More

Ferric Ammonium Citrate, Brown, Powder, FCC, 16.5-18.5%, Spectrum™ Chemical

Catalog No. p-7157416
Change view
Click to view available options
Quantity:
500 g
2.5 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602657 500 g Amber Glass Bottle
18602658 2.5 kg Poly Pail
18602659 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18602657 Supplier Spectrum Chemical Mfg Cor Supplier No. F1000500GM
Only null left

F1000, 1185-57-5

Spectrum™ Chemical Ferric Ammonium Citrate, Brown, Powder, FCC is a brownish powder which is easily soluble in water. It is a food additive used as an acidity regulator and anti-caking agent. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1185-57-5
Molecular Formula C6H8FeNO7
Molecular Weight (g/mol) 261.98
InChI Key FRHBOQMZUOWXQL-UHFFFAOYSA-K
IUPAC Name iron(3+) 2-hydroxypropane-1,2,3-tricarboxylate amine
SMILES N.[Fe+3].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

Specifications

CAS Max % 100%
Quantity 500 g
Percent Purity 16.5 to 18.5%
Grade FCC
Packaging Amber Glass Bottle