missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ T4 DNA Polymerase (5 U/μL)
Description
T4 DNA Polymerase, a template-dependent DNA polymerase, catalyzes 5'-3' synthesis from primed single-stranded DNA. The enzyme has a 3'-5' exonuclease activity, but lacks 5'-3' exonuclease activity.
Highlights
- Stronger 3'-5' exonuclease activity on single-stranded than on double-stranded DNA and greater (more than 200 times) than DNA polymerase I, E. coli,and Klenow fragment
- Active in Thermo Scientific restriction enzyme, PCR, RT and T4 DNA Ligase buffers
Specifications
Specifications
| Concentration | 5 U/μL |
| Polymerase | T4 DNA Polymerase |
| Quantity | 100 Units |
| Includes | 0.35 mL of 5X Reaction Buffer |
For Research Use Only. Not for use in diagnostic procedures.