Learn More
Thermo Scientific™ SP6 RNA Polymerase (20 U/μL)
Catalog No. FEREP0131
Change view
Click to view available options
Quantity:
2,000 units
Catalog No. Quantity
FEREP0131 2,000 units
1 options
Catalog No. FEREP0131 Supplier Thermo Scientific™ Supplier No. EP0131
Season of Savings
View all promotions
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Thermo Scientific™ SP6 RNA Polymerase is a DNA-dependent RNA polymerase with strict specificity for its double-stranded promoter.
Thermo Scientific™ Bacteriophage SP6 RNA polymerase is DNA-dependent RNA polymerase with strict specificity for their respective double-stranded promoters.
It catalyzes the 5'=>3' synthesis of RNA on either single-stranded DNA or double-stranded DNA downstream from it promoter.
  • Incorporates modified nucleotides (e.g., aminoallyl-, biotin-, fluorescein-, digoxigenin-labeled nucleotides)
Recommended for:
Synthesis of unlabeled and labeled RNA that can be used: for hybridization (1), in vitro RNA translation (2); as aRNA (3), siRNA (4), substrate in RNase protection assays (5), template for genomic DNA sequencing.
Note:
Consensus promoter sequences: AATTAACCCTCACTAAA G GGAGA - T3 TAATACGACTCACTATA G GGAGA - T7 ATTTAGGTGACACTATA G AAGNG - SP6 The position in bold (+1) indicates the first nucleotide incorporated into RNA during transcription. Only bases at this position through +3 are critical for transcription, and they must be a G and a purine base, respectively (9).
Specifications
Polymerase SP6 RNA Polymerase
Quantity 2,000 units
Concentration 1X
For Research Use Only. Not for use in diagnostic procedures.