Learn More
Thermo Scientific™ RiboRuler Low Range RNA Ladder, ready-to-use
Description
RiboRuler Low Range RNA Ladder 100-1000 bases, ready-to-use is a mixture of 7 chromatography purified single-stranded RNA transcripts (100-1000 bp long), produced from specific templates that contain a fragment of the pTZ19R polylinker and Lambda phage fragments.
This ladder is premixed with loading dye.
RiboRuler Low Range RNA Ladder, ready-to-use, is composed of chromatography-purified transcripts what makes them free from NTPs and RNA degradation products. The concentration of each RNA transcript is determined spectrophotometrically (see photo for precise quantities). To minimize the possibility of contamination during the use, the ladder is aliquotted in smaller volumes.
Highlights
- Sharp bands of uniform intensity
- Easy-to-remember band sizes and quantities
- Approximate RNA quantification by predetermined concentration of ladder bands
- Supplied with 2X RNA Loading Dye
- Stable for 6 months at -20°C
Specifications
Specifications
| Concentration | 2X |
| Gel Type | Agarose |
| Label or Dye | Xylene Cyanol FF, Bromophenol Blue, Ethidium Bromide |
| Size Range | 100 to 1000 bp |
| Quantity | 5 x 40 μL |
| Product Line | RiboRuler |
| Product Type | DNA Ladder |
For Research Use Only. Not for use in diagnostic procedures.