Learn More
Thermo Scientific™ RiboRuler High Range RNA Ladder, ready-to-use
Description
RiboRuler High Range RNA Ladder 200 to 6000 bases, ready-to-use is a mixture of 8 chromatography purified single-stranded RNA transcripts (200 to 6000 bp long), produced from specific templates that contain a fragment of the pTZ19R polylinker and Lambda phage fragments. Ladder is premixed with loading dye.
RiboRuler High Range RNA Ladder, ready-to-use, is composed of chromatography-purified transcripts that makes them free from NTPs and RNA degradation products. The concentration of each RNA transcript is determined spectrophotometrically (see photo for precise quantities). To minimize the possibility of contamination during the use, the ladder is aliquotted in smaller volumes.
Highlights
- Sharp bands of uniform intensity
- Easy-to-remember band sizes and quantities
- Predetermined concentration of ladder bands allows for approximate RNA quantification
- Supplied with 2X RNA Loading Dye
- Stable for 6 months at -20°C
Specifications
Specifications
| Concentration | 2X |
| Gel Type | Agarose |
| Label or Dye | Xylene Cyanol FF, Bromophenol Blue, Ethidium Bromide |
| Size Range | 200 to 6000 bp |
| Quantity | 5 x 40 μL |
| Product Line | RiboRuler |
| Product Type | High Range DNA Ladder |
For Research Use Only. Not for use in diagnostic procedures.