missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ RiboRuler Low Range RNA Ladder

Catalog No. fersm1831
Change view
Click to view available options
Quantity:
5 x 20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERSM1831 5 x 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERSM1831 Supplier Thermo Scientific™ Supplier No. SM1831
Only null left

Size and quantify RNA with Thermo Scientific™ RiboRuler Low Range RNA Ladder, a mixture of 7 chromatography purified single-stranded RNA transcripts (100 to 1000 bp long).

RiboRuler Low Range RNA Ladder 100-1000 bp is a mixture of 7 chromatography purified single-stranded RNA transcripts (100-1000 bp long), produced from specific templates that contain a fragment of the pTZ19R polylinker and Lambda phage fragments.

Ladder is available in 1 mM EDTA, pH 6.0.

RiboRuler Low Range RNA Ladder is composed of chromatography-purified transcripts what makes them free from NTPs and RNA degradation products. The concentration of each RNA transcript is determined spectrophotometrically (see Photo for precise quantities). To minimize the possibility of contamination during the use the ladder is aliquotted in smaller volumes.

RiboRuler Low Range RNA Ladder is optimal for end-labeling using T4 Polynucleotide Kinase and is ideal for Northern blots.

Highlights

  • Sharp bands of uniform intensity 
  • Easy-to-remember band sizes and quantities
  • Approximate RNA quantification by predetermined concentration of ladder bands
  • Supplied with 2X RNA Loading Dye 
  • Stable for 6 months at -20°C 

Specifications

Content And Storage • 5 x 20 μL RiboRuler Low Range RNA Ladder (in 1 mM EDTA, pH 6.0); sufficient for 50 uses
• 1 mL RNA Loading Dye

Store at -70°C or at -20°C (up to 6 months).

Concentration 2X
Gel Type Agarose
Label or Dye Xylene Cyanol FF, Bromophenol Blue, Ethidium Bromide
Size Range 100 to 1000 bp
Quantity 5 x 20 μL
Product Line RiboRuler
Product Type DNA Ladder

For Research Use Only. Not for use in diagnostic procedures.