Learn More
Thermo Scientific™ phiX174 DNA/BsuRI (HaeIII) Marker, ready-to-use
Description
0.5 μg/lane, 8cm length gel, 1X TBE, 5V/cm, 1.5h. Bands visible: 1353, 1078, 872, 603, 310 281, 271, 234, 194, 118, and 118 bp.
phiX174 DNA is digested to completion with the appropriate Thermo Scientific restriction enzyme, then purified and dissolved in storage buffer. The DNA fragments contain blunt or sticky ends, depending on the restriction enzyme used for the markers preparation.
phiX174 DNA/BsuRI (HaeIII) Marker, ready-to-use is premixed with 6X DNA Loading Dye for direct loading onto agarose and polyacrylamide gels.
Highlights
- Sizing and approximate quantification of small DNA fragments
- Sharp bands
- Premixed with 6X DNA Loading Dye for direct loading and room temperature storage
- Supplied with loading dye for sample DNA
Specifications
Specifications
| Concentration | 6X |
| Gel Type | Agarose |
| Ready to Load | Yes |
| Gel Compatibility | Agarose Gels |
| Quantity | 50 μg |
| Product Type | DNA/BSURI (HAEIII) Marker |
Frequently Asked Questions (FAQs)
Each of Thermo Scientific DNA and RNA ladders has a recommended voltage given in V/cm. Please refer to the gel picture in the product information sheet. V/cm stands for the amount of voltage recommended per cm of the distance between the two electrodes of the gel box (not gel length).
For Research Use Only. Not for use in diagnostic procedures.