missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Water, nuclease-free

Catalog No. FERR0582
Change view
Click to view available options
Quantity:
30 mL
4 x 1.25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERR0582 30 mL
FERR0581 4 x 1.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FERR0582 Supplier Thermo Scientific™ Supplier No. R0582

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Optimize molecular biology applications with this deionized and 0.22μm membrane-filtered nuclease-free water.

Nuclease-free water is deionized and 0.22 μm membrane-filtered nuclease-free water. It is ideal for all .

Ideal for all molecular biology applications.
  • Free of endo-, exodeoxyribonucleases, ribonucleases and phosphatases

Specifications

pH 7.6
Grade Molecular Biology
Packaging Bottle
Treatment(s) Not DEPC-Treated
Quantity 30 mL
I would like to order a TaqMan SNP Genotyping Assay. Does the kit include everything I need to perform my experiment or do I need to purchase reagents separately?

Each predesigned TaqMan SNP Genotyping Assay is delivered in a single tube consisting of two differentially labeled allele-specific TaqMan MGB (minor groove binding) probes and a pair of PCR primers. For more information, please see the following link: https://www.thermofisher.com/us/en/home/life-science/pcr/real-time-pcr/real-time-pcr-applications/genetic-variation-analysis-using-real-time/snp-genotyping-with-real-time-pcr.html

Besides the TaqMan SNP Genotyping Assay, you would need to purchase a Genotyping Master Mix and nuclease-free water. A list of compatible master mixes can be found in the following link: https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0009593_TaqManSNP_UG.pdf.

For Research Use Only. Not for use in diagnostic procedures.