Learn More
Thermo Scientific™ GeneRuler Ready-to-Use DNA Ladders
Thermo Scientific GeneRuler DNA Ladders offer a wide selection of size standard made with individual chromatography-purified fragments in the range of 10 bp to 48.5 kb.
$96.15 - $559.35
Specifications
| Concentration | 6X |
|---|---|
| Gel Type | Agarose |
| Ready to Load | Yes |
| Gel Compatibility | Agarose Gels |
| Product Line | GeneRuler |
| Catalog Number | Mfr. No. | Product Type | Quantity | Size Range | Number of Markers | Price | Quantity | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Product Type | Quantity | Size Range | Number of Markers | Price | Quantity | ||
|
FERSM1213
|
Thermo Scientific™
SM1213 |
Ultra Low Range DNA Ladder | 100 Applications | 10 to 300 bp | 11 |
N/A
|
|
||
|
FERSM1193
|
Thermo Scientific™
SM1193 |
Low Range DNA Ladder | 100 Applications | 25 to 700 bp | 10 |
N/A
|
|
||
|
FERSM0373
|
Thermo Scientific™
SM0373 |
50BP DNA Ladder | 100 Applications | 50 to 1000 bp | 13 |
N/A
|
|
||
|
FERSM1334
|
Thermo Scientific™
SM1334 |
1KB Plus DNA Ladder | 100 Applications | 75 to 20000 bp | 15 |
N/A
|
|
||
|
FERSM0243
|
Thermo Scientific™
SM0243 |
100BP DNA Ladder | 100 Applications | 100 to 1000 bp | 10 |
N/A
|
|
||
|
FERSM0323
|
Thermo Scientific™
SM0323 |
100BP Plus DNA Ladder | 100 Applications | 100 to 3000 bp | 14 |
N/A
|
|
||
|
FERSM0334
|
Thermo Scientific™
SM0334 |
DNA Ladder Mix | 100 Applications | 100 to 1000 bp | 21 |
N/A
|
|
||
|
FERSM0314
|
Thermo Scientific™
SM0314 |
1KB DNA Ladder | 100 Applications | 250 to 10000 bp | 14 |
N/A
|
|
||
|
FERSM1353
|
Thermo Scientific™
SM1353 |
High Range DNA Ladder | 100 Applications | 10171 to 48502 bp | 8 |
N/A
|
|
||
|
FERSM1333
|
Thermo Scientific™
SM1333 |
1KB Plus DNA Ladder | 500 Applications | 75 to 20000 bp | 15 |
N/A
|
|
||
|
FERSM0244
|
Thermo Scientific™
SM0244 |
100BP DNA Ladder | 500 Applications | 100 to 1000 bp | 10 |
N/A
|
|
||
|
FERSM0324
|
Thermo Scientific™
SM0324 |
100BP Plus DNA Ladder | 500 Applications | 100 to 3000 bp | 14 |
N/A
|
|
||
|
FERSM0333
|
Thermo Scientific™
SM0333 |
DNA Ladder Mix | 500 Applications | 100 to 1000 bp | 21 |
N/A
|
|
||
|
FERSM0313
|
Thermo Scientific™
SM0313 |
1KB DNA Ladder | 500 Applications | 250 to 10000 bp | 14 |
N/A
|
|
||
Description
Thermo Scientific GeneRuler DNA Ladders serve as precise molecular weight markers for the sizing and approximate quantification of double-stranded DNA fragments, accommodating a range from 10 bp up to 48.5 kb. The uniform intensity of the bands, coupled with distinct bright reference bands, facilitates accurate sizing and orientation during gel electrophoresis. Furthermore, the defined quantity of DNA within each band enables the approximate quantification of experimental DNA samples.
GeneRuler DNA ladders are provided in a convenient ready-to-use format incorporating 6X TriTrack DNA Loading Dye (contains Xylene Cyanol FF, Bromophenol Blue, and Orange G). The three dye present in the buffer allow for easy tracking of DNA migration during electrophoresis.
Highlights
- Wide selection of DNA ladders fitting a variety of experiments
- Sharp bands, easy-to-remember fragment sizes and quantities
- Easy visualization—three-colored loading dye for easy tracking of DNA migration during electrophoresis
- Ready-to-use: premixed with 6X MassRuler DNA Loading Dye for direct loading
- Stable at room temperature for up to 24 months
Sustainability features
Ready-to-use GeneRuler DNA ladders can be stored at room temperature for up to 24 months. The ladder is shipped at ambient temperature, using optimized right-sized packaging and 100% recyclable packaging reducing waste.
Specifications
| 6X | |
| Yes | |
| GeneRuler |
| Agarose | |
| Agarose Gels |
For Research Use Only. Not for use in diagnostic procedures.