missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneRuler Ready-to-Use DNA Ladders
GREENER_CHOICE

Catalog No. FERSM1334
Change view
Click to view available options
Product Type:
100BP DNA Ladder
100BP Plus DNA Ladder
1KB DNA Ladder
1KB Plus DNA Ladder
50BP DNA Ladder
DNA Ladder Mix
High Range DNA Ladder
Low Range DNA Ladder
Ultra Low Range DNA Ladder
Quantity:
100 Applications
500 Applications
Size Range:
10 to 300 bp
100 to 1000 bp
100 to 3000 bp
10171 to 48502 bp
25 to 700 bp
250 to 10000 bp
50 to 1000 bp
75 to 20000 bp
Number of Markers:
10
11
13
14
15
21
8
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Quantity Size Range Number of Markers
FERSM1334 1KB Plus DNA Ladder 100 Applications 75 to 20000 bp 15
FERSM1213 Ultra Low Range DNA Ladder 100 Applications 10 to 300 bp 11
FERSM1193 Low Range DNA Ladder 100 Applications 25 to 700 bp 10
FERSM0373 50BP DNA Ladder 100 Applications 50 to 1000 bp 13
FERSM0243 100BP DNA Ladder 100 Applications 100 to 1000 bp 10
FERSM0323 100BP Plus DNA Ladder 100 Applications 100 to 3000 bp 14
FERSM0334 DNA Ladder Mix 100 Applications 100 to 1000 bp 21
FERSM0314 1KB DNA Ladder 100 Applications 250 to 10000 bp 14
FERSM1353 High Range DNA Ladder 100 Applications 10171 to 48502 bp 8
FERSM1333 1KB Plus DNA Ladder 500 Applications 75 to 20000 bp 15
FERSM0244 100BP DNA Ladder 500 Applications 100 to 1000 bp 10
FERSM0324 100BP Plus DNA Ladder 500 Applications 100 to 3000 bp 14
FERSM0333 DNA Ladder Mix 500 Applications 100 to 1000 bp 21
FERSM0313 1KB DNA Ladder 500 Applications 250 to 10000 bp 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Catalog No. FERSM1334 Supplier Thermo Scientific™ Supplier No. SM1334
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific GeneRuler DNA Ladders offer a wide selection of size standard made with individual chromatography-purified fragments in the range of 10 bp to 48.5 kb.

Thermo Scientific GeneRuler DNA Ladders serve as precise molecular weight markers for the sizing and approximate quantification of double-stranded DNA fragments, accommodating a range from 10 bp up to 48.5 kb. The uniform intensity of the bands, coupled with distinct bright reference bands, facilitates accurate sizing and orientation during gel electrophoresis. Furthermore, the defined quantity of DNA within each band enables the approximate quantification of experimental DNA samples.

GeneRuler DNA ladders are provided in a convenient ready-to-use format incorporating 6X TriTrack DNA Loading Dye (contains Xylene Cyanol FF, Bromophenol Blue, and Orange G). The three dye present in the buffer allow for easy tracking of DNA migration during electrophoresis.

Highlights

  • Wide selection of DNA ladders fitting a variety of experiments
  • Sharp bands, easy-to-remember fragment sizes and quantities
  • Easy visualization—three-colored loading dye for easy tracking of DNA migration during electrophoresis
  • Ready-to-use: premixed with 6X MassRuler DNA Loading Dye for direct loading
  • Stable at room temperature for up to 24 months

Sustainability features

Ready-to-use GeneRuler DNA ladders can be stored at room temperature for up to 24 months. The ladder is shipped at ambient temperature, using optimized right-sized packaging and 100% recyclable packaging reducing waste.

Specifications

Concentration 6X
Gel Type Agarose
Label or Dye Xylene Cyanol FF, Bromophenol Blue, Orange G
Number of Markers 15
Ready to Load Yes
Size Range 75 to 20000 bp
Volume (Metric) 50 μg
Gel Compatibility Agarose Gels
Quantity 100 Applications
Product Line GeneRuler
Product Type 1KB Plus DNA Ladder
Show More Show Less
Are the Thermo Scientific DNA ladders compatible with GelRed and GelGreen gel stains?

The GelRed or GelGreen nucleic acid dyes can be used with our DNA Ladders. Nucleic acid binding dyes can affect DNA migration during electrophoresis, therefore we highly recommend post-staining of gels. Please follow the gel staining protocol provided by the manufacturer of nucleic acid gel dyes.

Which Thermo Scientific DNA ladders are optimal for fast electrophoresis separation?

For fast sizing and quantification of double-stranded DNA, we recommend using FastRuler DNA Ladders. These ladders are easily resolved in a short separation distance after an 8-14 min run.

What is the recommended voltage for electrophoresis with Thermo Scientific DNA and RNA ladders? What does V/cm mean?

Each of Thermo Scientific DNA and RNA ladders has a recommended voltage given in V/cm. Please refer to the gel picture in the product information sheet. V/cm stands for the amount of voltage recommended per cm of the distance between the two electrodes of the gel box (not gel length).

What are the end functional groups of Thermo Scientific DNA and RNA ladders?

Thermo Scientific DNA and RNA ladders have 5′-phosphate and 3′-hydroxyl groups.

For Research Use Only. Not for use in diagnostic procedures.