missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneRuler 50 bp DNA Ladder
GREENER_CHOICE

Catalog No. FERSM0372
Change view
Click to view available options
Quantity:
5 x 50 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERSM0372 5 x 50 μg
FERSM0371 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FERSM0372 Supplier Thermo Scientific™ Supplier No. SM0372
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Perform sizing and approximate quantification of a wide range of double-stranded DNA on agarose gels with this mixture of chromatography-purified individual DNA fragments.

GeneRuler 50bp DNA Ladder is recommended for sizing and for approximate quantification of a wide range of double-stranded DNA on agarose or polyacrylamide gels. The ladder is a mixture of chromatography-purified individual DNA fragments.

GeneRuler 50bp DNA Ladder can be labeled radioactively with T4Polynucleotide Kinase.

The ladder is supplied with 6X DNA Loading Dye.

The ladder is a mixture of chromatography-purified individual DNA fragments. GeneRuler 50 bp DNA Ladder can be labeled radioactively with T4 Polynucleotide Kinase. The ladder is supplied with 6X DNA Loading Dye.
  • Supplied with loading dye for sample DNA
  • Ideal for both DNA sizing and approximate quantification
  • Sharp bands
  • Bright reference bands (given in red)

Specifications

Concentration 6X
Gel Type Agarose
Label or Dye Xylene Cyanol FF, Bromophenol Blue, Orange G
Ready to Load No
Size Range 50 to 1000 bp
Gel Compatibility Agarose Gels
Quantity 5 x 50 μg
Product Line GeneRuler
Product Type DNA Ladder
Are the Thermo Scientific DNA ladders compatible with GelRed and GelGreen gel stains?

The GelRed or GelGreen nucleic acid dyes can be used with our DNA Ladders. Nucleic acid binding dyes can affect DNA migration during electrophoresis, therefore we highly recommend post-staining of gels. Please follow the gel staining protocol provided by the manufacturer of nucleic acid gel dyes.

Which Thermo Scientific DNA ladders are optimal for fast electrophoresis separation?

For fast sizing and quantification of double-stranded DNA, we recommend using FastRuler DNA Ladders. These ladders are easily resolved in a short separation distance after an 8-14 min run.

What is the recommended voltage for electrophoresis with Thermo Scientific DNA and RNA ladders? What does V/cm mean?

Each of Thermo Scientific DNA and RNA ladders has a recommended voltage given in V/cm. Please refer to the gel picture in the product information sheet. V/cm stands for the amount of voltage recommended per cm of the distance between the two electrodes of the gel box (not gel length).

What are the end functional groups of Thermo Scientific DNA and RNA ladders?

Thermo Scientific DNA and RNA ladders have 5′-phosphate and 3′-hydroxyl groups.

For Research Use Only. Not for use in diagnostic procedures.