missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ GeneRuler 100 bp DNA Ladder

Description
GeneRuler 100bp 100 to 1000bpDNA Ladder is recommended for sizing and approximate quantification of a wide range of double-stranded DNA on agarose or polyacrylamide gels. The ladder is a mixture of chromatography-purified individual DNA fragments.
GeneRuler 100bp DNA Ladder can be labeled radioactively with T4 Polynucleotide Kinase (seeProtocol).
The ladder is supplied with 6X DNA Loading Dye.
The ladder is a mixture of chromatography-purified individual DNA fragments. GeneRuler 100 bp DNA Ladder can be labeled radioactively with T4 Polynucleotide Kinase, protocol. The ladder is supplied with 6X DNA Loading Dye.
- Ideal for both DNA sizing and approximate quantification
- Sharp bands
- Bright reference bands (given in red)
- Supplied with loading dye for sample DNA
Specifications
Specifications
| Concentration | 6X |
| Gel Type | Agarose |
| Label or Dye | Xylene Cyanol FF, Bromophenol Blue, Orange G |
| Ready to Load | No |
| Size Range | 100 to 1000 bp |
| Gel Compatibility | Agarose Gels |
| Quantity | 50 μg |
| Product Line | GeneRuler |
| Product Type | DNA Ladder |
For Research Use Only. Not for use in diagnostic procedures.