missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ dNTP Set, 100 mM Solutions

Catalog No. FERR0181
Change view
Click to view available options
Quantity:
4 x 0.25 mL
4 x 1 mL
4 x 5 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERR0181 4 x 0.25 mL
FERR0182 4 x 1 mL
FERR0186 4 x 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. FERR0181 Supplier Thermo Scientific™ Supplier No. R0181
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific™ dNTP Set is a convenient set of 100 mM aqueous solutions of each of dATP, dCTP, dGTP and dTTP, supplied in separate vials.

Thermo Scientific™ dNTP Set is a convenient set of 100 mM aqueous solutions of each of dATP, dCTP, dGTP and dTTP, supplied in separate vials. These solutions of extremely stable nucleotides are titrated to pH 7.3-7.5 with NaOH. The nucleotides have greater than 99% purity, are free of nuclease activities, human and E. coli DNA. They are designed for many different molecular biology applications.

Features

  • Purity confirmed by HPLC
  • Free of human and E. coli DNA

Applications
For use in a wide variety of molecular biology applications, including:

  • High fidelity and Long range PCR (40 kb)
  • PCR, qPCR, RT-PCR, and cDNA synthesis
  • RDA, MDA, LAMP-PCR
  • DNA labeling and sequencing

Stablity

  • Stable after multiple freeze-thaw cycles
  • Up to 95% of dNTPs remain in triphosphate form after 7 weeks at room temperature
  • Up to 90% of dNTPs remain in triphosphate form after 30 cycles of PCR (1 min at 94°C, 3 min at 72°C)

Specifications

Concentration 100 mM
Content And Storage dNTP Set, 100 mM solutions (4 x 0.25 mL)

Store at –20°C.
Form Liquid
Label or Dye Unlabeled
Purity or Quality Grade >99%
Quantity 4 x 0.25 mL
Shipping Condition Dry Ice
For Use With (Application) Standard PCR, High-fidelity PCR, Real Time PCR (qPCR)
What buffer are the dNTP Set 100 mM Solutions stored in?

The buffer dNTP Set 100 mM Solutions are stored in is water (brought to the correct pH with NaOH).

For Research Use Only. Not for use in diagnostic procedures.