missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Aminoallyl-dUTP Solution (50 mM)

Catalog No. FERR1101
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERR1101 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERR1101 Supplier Thermo Scientific™ Supplier No. R1101

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific Aminoallyl-dUTP (5-[3-aminoallyl]-2'-deoxyuridine-5'-triphosphate) is supplied as 10 mM and 50 mM aqueous solutions titrated to pH 7.0 with NaOH.

Thermo Scientific Aminoallyl-dUTP (5-[3-aminoallyl]-2'-deoxyuridine-5'-triphosphate) is supplied as 10 mM and 50 mM aqueous solutions titrated to pH 7.0 with NaOH. Aminoallyl-dUTP can be enzymatically incorporated into DNA and the resulting amine-containing DNA can be subsequently labeled with any amine-reactive fluorescent dye, biotin or hapten.

Applications
• Enzymatic indirect non-radioactive labeling of DNA during cDNA synthesis
• PCR
• Nick-translation
• Random-primed labeling
• Primer extension

Aminoallyl-dUTP can be enzymatically incorporated into DNA with:
• Reverse transcriptases
• Taq DNA polymerase (Cat. No. EP0072)
• phi29 DNA Polymerase (EP0091)
• Klenow Fragment (EP0051)
• Klenow Fragment, exo- (EP0421)
• DNA Polymerase I (EP0041)

Specifications

Concentration 50 mM
Content And Storage Aminoallyl-dUTP Solution, 50 mM (50 μL)

Store at –20°C.
Form Liquid
Polymerase DNA Polymerase
Labeling Method Direct Labeling
Label or Dye Aminoallyl
Quantity 50 μL
Shipping Condition Dry Ice
For Use With (Application) Standard PCR, Gene Expression, Non-Radioactive Labeling

For Research Use Only. Not for use in diagnostic procedures.