missing translation for 'onlineSavingsMsg'
Learn More

Fenoprofen Calcium Salt Dihydrate 98.0+%, TCI America™

Catalog No. F037225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F037225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F037225G Supplier TCI America Supplier No. F037225G
Only null left

Chemical Identifiers

CAS 71720-56-4
Molecular Formula C30H32CaO8
Molecular Weight (g/mol) 560.66
MDL Number MFCD00242722,MFCD00079289
InChI Key LXVOQQSZQOHNFB-UHFFFAOYNA-N
Synonym 2-(3-Phenoxyphenyl)propionic Acid Calcium Salt
PubChem CID 131673867
IUPAC Name bis(2-(3-phenoxyphenyl)propanoic acid) calcium dihydrate
SMILES O.O.[Ca].CC(C(O)=O)C1=CC(OC2=CC=CC=C2)=CC=C1.CC(C(O)=O)C1=CC(OC2=CC=CC=C2)=CC=C1

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 522.61
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Fenoprofen Calcium Salt Dihydrate
RTECSNumber CY1678900
TSCA No