missing translation for 'onlineSavingsMsg'
Learn More

Fenclorim 98.0+%, TCI America™

Catalog No. p-7129939
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
F08511G 1 g
F08515G 5 g
2 options

Catalog No. F08511G

Supplier: TCI America F08511G

Only null left

Chemical Identifiers

CAS 3740-92-9
Molecular Formula C10H6Cl2N2
Molecular Weight (g/mol) 225.072
MDL Number MFCD00754193
InChI Key NRFQZTCQAYEXEE-UHFFFAOYSA-N
Synonym 4,6-Dichloro-2-phenylpyrimidine
PubChem CID 77338
ChEBI CHEBI:81993
IUPAC Name 4,6-dichloro-2-phenylpyrimidine
SMILES C1=CC=C(C=C1)C2=NC(=CC(=N2)Cl)Cl

Specifications

Melting Point 97°C
Color Yellow
Quantity 1 g
Formula Weight 225.07
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Fenclorim
EINECSNumber (5)-5620
RTECSNumber UV8258400
TSCA No