missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fenclorim 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 97°C |
| Color | Yellow |
| Quantity | 5 g |
| Formula Weight | 225.07 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Fenclorim |
Safety and Handling
| EINECSNumber | (5)-5620 |
| RTECSNumber | UV8258400 |
| TSCA | No |