missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(-)-Fenchone 98.0+%, TCI America™
$55.07 - $394.92
Chemical Identifiers
| CAS | 7787-20-4 |
|---|---|
| Molecular Formula | C10H16O |
| Molecular Weight (g/mol) | 152.237 |
| MDL Number | MFCD00151104 |
| InChI Key | LHXDLQBQYFFVNW-GMSGAONNSA-N |
| Synonym | l-fenchone, 1r-fenchone, 1r,4r-2,2,4-trimethylbicyclo 2.2.1 heptan-3-one, 2r,5r-2,6,6-trimethyl-2,5-methanocyclohexane-1-one |
| PubChem CID | 3034206 |
| IUPAC Name | (1R,4R)-2,2,4-trimethylbicyclo[2.2.1]heptan-3-one |
| SMILES | CC1(C2CCC(C2)(C1=O)C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
F016425ML
|
TCI America
F016425ML |
25 mL |
Each for $55.07
|
|
|||||
|
F0164500ML
|
TCI America
F0164500ML |
500 mL |
Each for $394.92
|
|
|||||
Chemical Identifiers
| 7787-20-4 | |
| 152.237 | |
| LHXDLQBQYFFVNW-GMSGAONNSA-N | |
| 3034206 | |
| CC1(C2CCC(C2)(C1=O)C)C |
| C10H16O | |
| MFCD00151104 | |
| l-fenchone, 1r-fenchone, 1r,4r-2,2,4-trimethylbicyclo 2.2.1 heptan-3-one, 2r,5r-2,6,6-trimethyl-2,5-methanocyclohexane-1-one | |
| (1R,4R)-2,2,4-trimethylbicyclo[2.2.1]heptan-3-one |
Specifications
| 7787-20-4 | |
| 193°C | |
| MFCD00151104 | |
| 1224 | |
| LHXDLQBQYFFVNW-GMSGAONNSA-N | |
| (1R,4R)-2,2,4-trimethylbicyclo[2.2.1]heptan-3-one | |
| 3034206 | |
| ≥98.0% (GC) | |
| (-)-Fenchone |
| Yellow | |
| C10H16O | |
| 25 mL | |
| l-fenchone, 1r-fenchone, 1r,4r-2,2,4-trimethylbicyclo 2.2.1 heptan-3-one, 2r,5r-2,6,6-trimethyl-2,5-methanocyclohexane-1-one | |
| CC1(C2CCC(C2)(C1=O)C)C | |
| 152.237 | |
| 152.24 | |
| Liquid |
Safety and Handling
EINECSNumber : (4)-1978
TSCA : Yes