missing translation for 'onlineSavingsMsg'
Learn More

(-)-Fenchone 98.0+%, TCI America™

Catalog No. F0164500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0164500ML 500 mL
F016425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F0164500ML Supplier TCI America Supplier No. F0164500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7787-20-4
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.237
MDL Number MFCD00151104
InChI Key LHXDLQBQYFFVNW-GMSGAONNSA-N
Synonym l-fenchone, 1r-fenchone, 1r,4r-2,2,4-trimethylbicyclo 2.2.1 heptan-3-one, 2r,5r-2,6,6-trimethyl-2,5-methanocyclohexane-1-one
PubChem CID 3034206
IUPAC Name (1R,4R)-2,2,4-trimethylbicyclo[2.2.1]heptan-3-one
SMILES CC1(C2CCC(C2)(C1=O)C)C

Specifications

Color Yellow
Boiling Point 193°C
Quantity 500 mL
UN Number 1224
Formula Weight 152.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (-)-Fenchone
EINECSNumber (4)-1978
TSCA Yes